Alfa Aesar™ Palladium, 0.5% on 3.18mm (0.125in) alumina pellets, unreduced

Manufacturer: Alfa Aesar™ 8911436

Quantity 500g

 View more versions of this product

Catalog No. AA8911436

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD03613602
    Chemical Name or Material Palladium, 0.5% on alumina pellets, 3.18mm (0.125 in.), Unreduced

    Specifications

    TSCA TSCA
    MDL Number MFCD03613602
    Quantity 500g
    Chemical Name or Material Palladium, 0.5% on alumina pellets, 3.18mm (0.125 in.), Unreduced

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit