Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium on 1/8-inch alumina pellets, 0.5% Pd, ACROS Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
7440-05-3
Pd
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3
Pd
106.42
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
Additional Information Loss on abrasion: 3.5%
Quantity 50g
Density 700g/L (tapped)
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Loss on Drying 1% (typically)
Packaging Glass bottle
Formula Weight 106.42
Physical Form Pellets
Percent Purity 0.5% (Pd)
Pore Volume 0.54mL/g
Specific Surface Area 230m2/g
Color Gray
Chemical Name or Material Palladium on 1/8-inch alumina pellets
Show More Show Less