Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium, 10% on activated carbon paste, 10R394, Thermo Scientific Chemicals

Catalog Number p-7059885
Click to view available options
Quantity:
5 g
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3
Pd
106.42
MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]

Palladium is used as a catalyst in the Suzuki reaction, hydrogenation reactions, deoxygenation, oxidation reactions, coupling reactions and Stille reaction. It is also used in jewelry, dentistry, watch making, blood sugar test strips, aircraft spark plugs, in the production of surgical instruments and electrical contacts and to make professional transverse flutes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Black
Quantity 5 g
Solubility Information Soluble in aqua regia. Insoluble in water.
Odor Odorless
Chemical Name or Material Palladium, 10% on activated carbon paste, 10R394
EINECSNumber 231-115-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit