Palladium, 10% on activated carbon powder, standard, reduced, Alfa Aesar™

Manufacturer: Alfa Aesar 4551518

Quantity 50g

 View more versions of this product

Catalog No. AA4551518

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    Physical Form Powder
    Molecular Formula Fe:Cr:Si:C;82.9:16.5:0.5:0.02wt%
    MDL Number MFCD03457879
    Chemical Name or Material Palladium, 10% on activated carbon powder

    Specifications

    Quantity 50g
    Color Dark gray to black
    DOT Information DOT Class: 4.1, Packaging Group: III
    Physical Form Powder
    Health Hazard 1 H228: Flammable solid.
    Appearance Dark gray to black powder
    Molecular Formula Fe:Cr:Si:C;82.9:16.5:0.5:0.02wt%
    MDL Number MFCD03457879
    Odor Odorless
    Chemical Name or Material Palladium, 10% on activated carbon powder

    H228: Flammable solid.

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit