Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Palladium, 10% on carbon, Type 487, dry

Manufacturer:  Alfa Aesar™ A1201218

Quantity 50g

 View more versions of this product

Catalog No. AAA1201218


  Check Availability
Add to cart

Specifications

Description

Applications
It is a catalyst used in Stille reaction, hydrogenation reactions, Suzuki-Miyaura cross-coupling and Heck-Mizoroki reactions, deoxygenation, oxidation reactions and coupling reactions.

Solubility
Insoluble in water and organic solvents.

Notes
Palladium on carbon should always be handled under an inert atmosphere (preferably argon), and reaction vessels should be flushed with inert gas before the catalyst is added.
Description & Specifications

Chemical Identifiers

7440-05-3
UN1325
Palladium, 10% on carbon, Type 487, Dry
MFCD00011167
14,6989

Specifications

7440-05-3
UN1325
50g
MFCD00011167
14,6989
Palladium, 10% on carbon, Type 487, Dry

Safety and Handling

SDS & Safety Documents

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, powder or water spray.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit