Alfa Aesar™ Palladium, 3% on activated carbon powder, eggshell, reduced

Manufacturer: Alfa Aesar™ 1169106

Quantity 5g

 View more versions of this product

Catalog No. AA1169106

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD03457879
    UN Number UN3178
    Chemical Name or Material Palladium, 3% on activated carbon powder, Eggshell, Reduced

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Health Hazard 1 Warning
    Odor Odorless
    Weight 900 to 1100m2/g
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
    TSCA TSCA
    MDL Number MFCD03457879
    UN Number UN3178
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    Quantity 5g
    Chemical Name or Material Palladium, 3% on activated carbon powder, Eggshell, Reduced

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit