Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Palladium, 3% on activated carbon powder, standard, reduced, nominally 50% water wet

Click to view available options
Quantity:
5g
25g
100g
7440-05-3
Pd
106.42
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3
Pd
106.42
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
Physical Form 900-1100m
Odor Odorless
Quantity 25g
Weight 900 to 1100m2/g
Chemical Name or Material Palladium, 3% on activated carbon powder, Standard, reduced, nominally 50% water wet

P210-P240-P241-P280-P370+P378q

H228

TSCA : Yes

Recommended Storage : Ambient temperatures