Alfa Aesar™ Palladium, 4.5% Rhodium, 0.5% on carbon paste, F101023-4.5/0.5

Manufacturer: Alfa Aesar™ H3716914

Quantity 25g

 View more versions of this product

Catalog No. AAH3716914

  Check Availability
Add to Cart

    Chemical Identifiers

    Chemical Name or Material Palladium, 4.5% Rhodium, 0.5% on carbon paste, F101023-4.5

    Specifications

    Physical Form Solid
    Color Dark gray
    Quantity 25g
    Odor Odorless
    Chemical Name or Material Palladium, 4.5% Rhodium, 0.5% on carbon paste, F101023-4.5

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit