Alfa Aesar™ Palladium, 5% on activated carbon paste, 5R437

Manufacturer: Alfa Aesar™ H3353718

Quantity 50g

 View more versions of this product

Catalog No. AAH3353718

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-05-3
    MDL Number MFCD03457879
    UN Number UN1325
    Chemical Name or Material Palladium, 5% on activated carbon paste, 5R437

    Specifications

    TSCA TSCA
    MDL Number MFCD03457879
    CAS 7440-05-3
    UN Number UN1325
    Quantity 50g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
    Chemical Name or Material Palladium, 5% on activated carbon paste, 5R437

    Description

    Applications
    It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

    Solubility
    Sparingly soluble in water.(0.26 g/L) (25°C),

    Notes
    Sold on a dry weight basis. Unit weight excludes water weight

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit