Alfa Aesar™ Palladium, 5% on activated carbon powder, standard, reduced, acidic catalyst, nominally 50% water wet

Manufacturer: Alfa Aesar™ 4414209

Quantity 10g

 View more versions of this product

Catalog No. AA4414209

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD03457879
    Chemical Name or Material Palladium, 5% on activated carbon powder, Standard, Reduced, Acidic catalyst, Nominally 50% water wet

    Specifications

    TSCA TSCA
    Physical Form Powder
    Quantity 10g
    Surface Area 900 to 1100m2/g
    MDL Number MFCD03457879
    Odor Odorless
    Chemical Name or Material Palladium, 5% on activated carbon powder, Standard, Reduced, Acidic catalyst, Nominally 50% water wet

    Description

    • Hydrogenation of aromatic and aliphatic nitro groups, reductive alkylation/amination, hydrogenation of aromatic carbonyls, olefins, nitriles

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit