Alfa Aesar™ Palladium, 5% on activated carbon powder, Type 487

Manufacturer: Alfa Aesar™ H3758722

Quantity 100g

 View more versions of this product

Catalog No. AAH3758722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-05-3
    MDL Number MFCD03457879
    UN Number UN1362
    Chemical Name or Material Palladium, 5% on activated carbon powder, Type 487

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P280-P235+P410-P240-P370+P378a-P420
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep cool.
    Protect from sunlight.
    Ground/bond container and receiving equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store away from other materials.
    TSCA TSCA
    Physical Form Powder
    UN Number UN1362
    Health Hazard 2 GHS H Statement
    H228-H252
    Flammable solid.
    Self-heating in large quantities
    may catch fire.
    Odor Odorless
    MDL Number MFCD03457879
    CAS 7440-05-3
    Quantity 100g
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 4.2; Packaging Group: III; Proper Shipping Name: CARBON, ACTIVATED
    Color Dark gray
    Chemical Name or Material Palladium, 5% on activated carbon powder, Type 487

    GHS H Statement
    H228-H252
    Flammable solid.
    Self-heating in large quantities
    may catch fire.

    GHS P Statement
    P210-P280-P235+P410-P240-P370+P378a-P420
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Keep cool.
    Protect from sunlight.
    Ground/bond container and receiving equipment.
    In case of fire: Use for extinction: CO2, powder or water spray.
    Store away from other materials.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit