Learn More
Alfa Aesar™ Palladium, 5% on activated carbon powder, Type A401102-5, eggshell unreduced, nominally 50% water wet
Aromatic nitro group hydrogenation
Manufacturer:  Alfa Aesar™ 3829922
Quantity 100g
 View more versions of this product
Catalog No. AA3829922
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
MFCD03457879
Palladium, 5% on activated carbon powder, Type A401102-5, eggshell unreduced, nominally 50% water wet
Specifications
MFCD03457879
100g
900 to 1100m2/g
Black
Odorless
Palladium, 5% on activated carbon powder, Type A401102-5, eggshell unreduced, nominally 50% water wet
Safety and Handling
SDS & Safety Documents
TSCA : TSCA