Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Palladium, 5% on barium sulfate, A308053-5

Manufacturer:  Alfa Aesar™ H3657522

Quantity 100g

 View more versions of this product

Catalog No. AAH3657522


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7440-05-3
MFCD03613605
Palladium, 5% on barium sulfate, A308053-5

Specifications

7440-05-3
MFCD03613605
Black
Palladium, 5% on barium sulfate, A308053-5
Powder or Pellets
Odorless
100g

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit