Alfa Aesar™ Palladium, 5% on calcium carbonate, A303060-5

Manufacturer: Alfa Aesar™ H3662214

Quantity 25g

 View more versions of this product

Catalog No. AAH3662214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-05-3
    MDL Number MFCD03427452
    Chemical Name or Material Palladium, 5% on calcium carbonate, A303060-5

    Specifications

    TSCA TSCA
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD03427452
    CAS 7440-05-3
    Color Dark brown or gray to black
    Quantity 25g
    Chemical Name or Material Palladium, 5% on calcium carbonate, A303060-5

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit