Alfa Aesar™ Palladium, 5% on calcium carbonate, lead poisoned, A305060-5

Manufacturer: Alfa Aesar™ H3618614

Quantity 25g

 View more versions of this product

Catalog No. AAH3618614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-05-3
    MDL Number MFCD03427452
    Synonym Lindlar catalyst
    Chemical Name or Material Palladium, 5% on calcium carbonate, lead poisoned, A305060-5

    Specifications

    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Physical Form Powder
    MDL Number MFCD03427452
    CAS 7440-05-3
    Synonym Lindlar catalyst
    Color Dark gray
    Health Hazard 1 Danger
    TSCA TSCA
    Quantity 25g
    Health Hazard 2 GHS H Statement
    H360-H201
    May damage fertility or the unborn child.
    Explosive
    mass explosion hazard.
    Chemical Name or Material Palladium, 5% on calcium carbonate, lead poisoned, A305060-5

    GHS H Statement
    H360-H201
    May damage fertility or the unborn child.
    Explosive
    mass explosion hazard.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit