Alfa Aesar™ Palladium, 5% on carbon paste, Type 761

Manufacturer: Alfa Aesar™ H3752622

Quantity 100g

 View more versions of this product

Catalog No. AAH3752622

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-05-3
    MDL Number MFCD03457879
    Chemical Name or Material Palladium, 5% on carbon paste, Type 761

    Specifications

    Odor Odorless
    TSCA TSCA
    Physical Form Solid
    MDL Number MFCD03457879
    Color Dark gray
    CAS 7440-05-3
    Quantity 100g
    Chemical Name or Material Palladium, 5% on carbon paste, Type 761

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit