Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium on activated carbon, 10% Pd, unreduced, dry, ACROS Organics™

Click to view available options
Quantity:
1g
50g
250g
Packaging:
Glass Bottle
7440-05-3,7440-44-0
Pd
106.42
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3,7440-44-0
Pd
106.42
MFCD03457879
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
Quantity 250g
UN Number 3178
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Merck Index 15, 7090
Packaging Glass Bottle
Formula Weight 106.42
Physical Form Powder
Percent Purity 10% (Pd)
Particle Size Particle size distribution: ≤4 Micron: 10%, ≤21 Micron: 50%, ≤90 Micron: 90%
Specific Surface Area 800m2/g
Color Black
Chemical Name or Material Palladium on Activated Carbon, 10% Pd
Show More Show Less

GHS H Statement:
Flammable solid.

GHS P Statement:
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning