Palladium anchored homogeneous catalyst, Alfa Aesar™, FibreCat™ 1007

For Chloro Coupling

Manufacturer: Alfa Aesar 44110-06

Quantity 5g

 View more versions of this product

Catalog No. AA4411006

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    Physical Form Orange fibers
    MDL Number MFCD04040827
    Chemical Name or Material Palladium anchored homogeneous catalyst

    Specifications

    Appearance Orange fibers
    Odor Odorless
    Color Orange
    Quantity 5g
    Physical Form Orange fibers
    MDL Number MFCD04040827
    Chemical Name or Material Palladium anchored homogeneous catalyst

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit