Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium on barium sulfate, 5% Pd, unreduced, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7440-05-3
Pd
106.42
MFCD03613605
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3
Pd
106.42
MFCD03613605
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
Quantity 10g
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Merck Index 15, 7090
Packaging Glass bottle
Formula Weight 106.42
Physical Form Powder
Percent Purity 4.50 to 5.50% (Pd)
Water 5% Max. (K.F.)
Color Brown
Chemical Name or Material Palladium on barium sulfate, 5% Pd

EINECSNumber : 231-115-6

RTECSNumber : RT3480500

TSCA : TSCA