Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium black, 98% Pd, surface area 28-33 m2/g, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
7440-05-3
Pd
106.42
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3
Pd
106.42
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
Quantity 5g
Calcium (Ca) 25ppm max.
Copper (Cu) 25ppm max.
Gold (Au) 50ppm max.
Linear Formula Pd
Iridium (Ir) 50ppm max.
Iron (Fe) 25ppm max.
Lead (Pb) 25ppm max.
Merck Index 15, 7090
Magnesium (Mg) 25ppm max.
Packaging Glass bottle
Formula Weight 106.4
Physical Form Powder
Percent Purity ≥97.5% (Pd)
Platinum (Pt) 50ppm max.
Rhodium (Rh) 50ppm max.
Ruthenium (Ru) 50ppm max.
Silicon (Si) 50ppm max.
Silver (Ag) 50ppm max.
Color Black
Melting Point 1555.0°C
Boiling Point 2970.0°C
Chemical Name or Material Palladium black, 98% Pd
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable solid.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 231-115-6

RTECSNumber : RT3480500

TSCA : TSCA