Palladium black, 99.9% (metals basis), Alfa Aesar™

Use as a catalyst in hydrogenation reactions. Other uses include the manufacture of paints, adhesives, fibers and coatings. Basic particles of Palladium black have the tendency to cluster into macroscopic aggregates.

Manufacturer: Alfa Aesar 0065906

Quantity 5g

 View more versions of this product

Catalog No. AA0065906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-05-3
    Physical Form Powder
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00011167
    Chemical Name or Material Palladium black

    Specifications

    Melting Point 1555°C
    Quantity 5g
    Solubility Information Insoluble in water. Soluble in aqua regia and fused alkalis.
    Color Gray to silver
    DOT Information DOT Class: 4.1, Packaging Group: II
    Density 12.02g/cm3 at 20°C
    EINECS Number 231-115-6
    Physical Form Powder
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00011167
    Odor Odorless
    Appearance Powder
    CAS 7440-05-3
    Health Hazard 1 H228: Flammable solid.
    Chemical Name or Material Palladium black

    H228: Flammable solid.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit