Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium on calcium carbonate, poisoned with 3.5% lead, 5% Pd, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7440-05-3,1314-08-5,7439-92-1,598-63-0,1317-36-8,471-34-1
Pd
106.42
MFCD00151707
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3,1314-08-5,7439-92-1,598-63-0,1317-36-8,471-34-1
Pd
106.42
MFCD00151707
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
Additional Information poisoned with 3.5% lead
Quantity 50g
UN Number 3077
Fieser 01,778; 04,365; 05,498; 06,443; 07,274; 15,13; 16,356
Merck Index 15, 7090
Packaging Glass bottle
Formula Weight 106.42
Physical Form Powder
Percent Purity 5% (Pd)
Color Black-Gray to Gray
Chemical Name or Material Palladium on calcium carbonate
Show More Show Less

GHS P Statement
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Harmful if swallowed.
May damage the unborn child. Suspected of damaging fertility.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF exposed or concerned: Get medical attention/advice.
Avoid release to the environment. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Danger

EINECSNumber : 231-115-6

RTECSNumber : RT3480500

TSCA : TSCA