Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium on calcium carbonate, reduced 5% Pd, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
7440-05-3,471-34-1
Pd
106.42
MFCD00151707
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3,471-34-1
Pd
106.42
MFCD00151707
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
Quantity 10g
Merck Index 15, 7090
Packaging Glass bottle
Palladium (Pd) 5%, exact Pd-content on the certificate of analysis
Formula Weight 106.42
Physical Form Powder
Water 5% Max.
Color Gray
Chemical Name or Material Palladium on calcium carbonate

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.