Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Palladium foil, 0.5mm (0.02 in.) thick, 99.95+% (metals basis)

Click to view available options
Quantity:
25 x 25mm
50 x 50mm
7440-05-3
Pd
106.42
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
7440-05-3
Pd
106.42
MFCD00011167
KDLHZDBZIXYQEI-UHFFFAOYSA-N
black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon
23938
CHEBI:33363
palladium
[Pd]
Quantity 50 x 50mm
Weight ≈3.8g/25x25mm
Odor Odorless
Physical Form Foil
Percent Purity ≥99.95%
Chemical Name or Material Palladium foil, 0.5mm (0.02 in.) thick
Assay (metals basis)

EINECSNumber : 231-115-6

TSCA : Yes

Recommended Storage : Ambient temperatures