Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Palladium hydroxide, 99%, Alfa Aesar™
Click to view available options
Quantity:
0.25g
1g
5g
Specifications
Assay Percent Range | 99% |
Solubility Information | Insoluble in water. |
Formula Weight | 140.42 |
Physical Form | Powder |
Percent Purity | 99% |
Odor | Odorless |
Color | Black |
Quantity | 5g |
Chemical Name or Material | Palladium hydroxide |
Safety and Handling
EINECSNumber : 235-219-2
TSCA : Yes
Recommended Storage : Ambient temperatures