Alfa Aesar™ Palladium(II) 2,4-pentanedionate, Pd 34.7%

Carbonylation, oligomerization

Manufacturer: Alfa Aesar™ 1051703

Quantity 1g

 View more versions of this product

Catalog No. AA1051703

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14024-61-4
    Formula Weight 304.92
    MDL Number MFCD00000025
    Synonym Palladium(II) acetylacetonate; Bis(acetylacetonato)palladium(II)
    Chemical Name or Material Palladium(II) 2,4-pentanedionate

    Specifications

    Formula Weight 304.92
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    MDL Number MFCD00000025
    Health Hazard 2 GHS H Statement
    H318
    Causes serious eye damage.
    TSCA TSCA
    Odor Odorless
    Physical Form Powder
    CAS 14024-61-4
    Synonym Palladium(II) acetylacetonate; Bis(acetylacetonato)palladium(II)
    Color Yellow to orange
    Solubility Information Soluble in benzene and chloroform
    Health Hazard 1 Danger
    Concentration Pd 34.7%
    Melting Point 210°C (dec.)
    Quantity 1g
    Chemical Name or Material Palladium(II) 2,4-pentanedionate

    GHS H Statement
    H318
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit