Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) acetate, 47.5% Pd, ACROS Organics™

Click to view available options
Quantity:
1g
2g
10g
50g
Packaging:
Glass bottle
3375-31-3
C4H6O4Pd
224.51
MFCD00012453
YJVFFLUZDVXJQI-UHFFFAOYSA-L
palladium ii acetate,palladium diacetate,palladium acetate,diacetoxypalladium,bis acetato palladium,bisacetylpalladium,diacetatopalladium,palladium 2+ acetate,palladous acetate
167845
palladium(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Pd+2]
3375-31-3
C4H6O4Pd
224.51
MFCD00012453
YJVFFLUZDVXJQI-UHFFFAOYSA-L
palladium ii acetate,palladium diacetate,palladium acetate,diacetoxypalladium,bis acetato palladium,bisacetylpalladium,diacetatopalladium,palladium 2+ acetate,palladous acetate
167845
palladium(2+);diacetate
CC(=O)[O-].CC(=O)[O-].[Pd+2]
Linear Formula (CH3CO2)2Pd
Merck Index 15,7092
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,methylene dichloride,,chloroform,acetonitrile and diethylether,insoluble in alcohols
Formula Weight 224.51
Percent Purity 47.5% Pd
Chloride 1000ppm max
Packaging Glass bottle
Solubility Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols
Melting Point 205.0°C
Quantity 10g
Chemical Name or Material Palladium(II) acetate, 47.5% Pd
Show More Show Less

GHS H Statement
Causes serious eye damage.
May cause long lasting harmful effects to aquatic life.

GHS P Statement
Wear eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 222-164-4

RTECSNumber : AJ1900000

TSCA : TSCA