Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) acetylacetonate, 35% Pd, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
14024-61-4
C10H14O4Pd
304.61
MFCD00000025
JKDRQYIYVJVOPF-SYWGCQIGSA-L
palladium diacetonate,acetylacetone palladium ii salt,bis 2,4-pentanedionato palladium ii
53384484
(E)-4-oxopent-2-en-2-olate;palladium(2+)
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Pd+2]
14024-61-4
C10H14O4Pd
304.61
MFCD00000025
JKDRQYIYVJVOPF-SYWGCQIGSA-L
palladium diacetonate,acetylacetone palladium ii salt,bis 2,4-pentanedionato palladium ii
53384484
(E)-4-oxopent-2-en-2-olate;palladium(2+)
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].[Pd+2]
Linear Formula (CH3COCH=C(CH3)O)2Pd
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in benzene
Formula Weight 304.61
Physical Form Solution
Percent Purity 35% Pd
Infrared Spectrum Authentic
Packaging Glass bottle
Solubility Solubility in water: insoluble in water. Other solubilities: soluble in benzene
Color Brown-Red
Melting Point 190.0°C
Quantity 5g
Chemical Name or Material Palladium(II) acetylacetonate, 35% Pd
Show More Show Less

GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc

GHS Signal Word: Danger

EINECSNumber : 237-859-8

TSCA : TSCA