Alfa Aesar™ Palladium(II) chloride, Premion™, 99.999% (metals basis), Pd 59.5% min

Manufacturer: Alfa Aesar™ 4308514

Quantity 25g

 View more versions of this product

Catalog No. AA4308514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-10-1
    Molecular Formula PdCl2
    Formula Weight 177.31
    MDL Number MFCD00003558
    Merck Index 14,6990
    Chemical Name or Material Palladium (II) chloride
    Grade Premion™

    Specifications

    Formula Weight 177.31
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P310-P406-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    tore in corrosive resistant/... container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Molecular Formula PdCl2
    Health Hazard 2 GHS H Statement
    H318-H290-H317
    Causes serious eye damage.
    May be corrosive to metals.
    May cause an allergic skin reaction.
    Melting Point 675°C (dec.)
    Quantity 25g
    TSCA TSCA
    MDL Number MFCD00003558
    Merck Index 14,6990
    Color Red-brown
    Density 4.0g/mL
    Physical Form Powder
    CAS 7647-10-1
    Concentration Pd 59.5% min
    RTECS Number RT3500000
    Health Hazard 1 Danger
    Chemical Name or Material Palladium (II) chloride
    Assay 99.999%, (metals basis)
    Grade Premion™

    GHS H Statement
    H318-H290-H317
    Causes serious eye damage.
    May be corrosive to metals.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P305+P351+P338-P310-P406-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    tore in corrosive resistant/... container with a resistant inner liner.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit