Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium (II) Chloride Solution, 5% w/v (Certified), Fisher Chemical

Click to view available options
Quantity:
100mL
Packaging:
Glass Bottle
7648-01-3,7732-18-5,7647-01-0
C5H7NOS2
161.24
MFCD00003558
UPCYEFFISUGBRW-UHFFFAOYSA-N
3-ethylrhodanine,n-ethylrhodanine,4-thiazolidinone, 3-ethyl-2-thioxo,rhodanine, 3-ethyl,3-ethyl-2-thioxothiazolidin-4-one,3-ethylrodanin,3-ethyl-2-thioxo-4-thiazolidinone,3-ethylrhodamine,3-ethyl-2-thioxo-1,3-thiazolidin-4-one,unii-42klq911ht
82098
3-ethyl-2-sulfanylidene-1,3-thiazolidin-4-one
CCN1C(=O)CSC1=S
7648-01-3,7732-18-5,7647-01-0
C5H7NOS2
161.24
MFCD00003558
UPCYEFFISUGBRW-UHFFFAOYSA-N
3-ethylrhodanine,n-ethylrhodanine,4-thiazolidinone, 3-ethyl-2-thioxo,rhodanine, 3-ethyl,3-ethyl-2-thioxothiazolidin-4-one,3-ethylrodanin,3-ethyl-2-thioxo-4-thiazolidinone,3-ethylrhodamine,3-ethyl-2-thioxo-1,3-thiazolidin-4-one,unii-42klq911ht
82098
3-ethyl-2-sulfanylidene-1,3-thiazolidin-4-one
CCN1C(=O)CSC1=S

In dilute HCl

Assay Percent Range 5% w/v
Density 1.0g/cm³
Concentration or Composition (by Analyte or Components) 5% w/v
Physical Form Liquid
Percent Purity 4.9 to 5.1%
Grade Certified
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 14mmHg at 20°C
Boiling Point 100°C
Color Yellow
Melting Point 0°C
pH 2
Quantity 100mL
Chemical Name or Material Palladium (II) Chloride Solution, 5% w/v
Show More Show Less