Alfa Aesar™ Palladium(II) iodide, Premion™, 99.998% (metals basis), Pd 29% min

Manufacturer: Alfa Aesar™ 43084-09

Quantity 10g

 View more versions of this product

Catalog No. AA4308409

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7790-38-7
    Molecular Formula PdI2
    Formula Weight 360.23g/mol
    MDL Number MFCD00011171
    Chemical Name or Material Palladium (II) iodide
    Grade Premion™

    Specifications

    Concentration Pd 29% min
    Density 6.003g/mL
    Color Black
    Melting Point 350°C (dec.)
    Quantity 10g
    CAS 7790-38-7
    Physical Form Powder
    Molecular Formula PdI2
    Formula Weight 360.23g/mol
    MDL Number MFCD00011171
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H317
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Chemical Name or Material Palladium (II) iodide
    Assay 99.998%, (metals basis)
    Grade Premion™

    GHS H Statement
    H317
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P363-P321-P333+P313-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash contaminated clothing before reuse.
    Specific treatment (see label).
    If skin irritation or rash occurs: Get medical advice/attention.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit