Learn More
Palladium(II) nitrate hydrate, 99.8% (metals basis), Pd 39% min, Thermo Scientific Chemicals
Description
Palladium(II) nitrate hydrate is used as a precursor to supported palladium catalysts. It is also used as a catalyst in the conversion of alkene nitration to glycol dinitrates. It is involved in the separation of chlorine and iodine and used as biochemical for proteomics research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | Red to Brown |
Physical Form | Crystals |
Quantity | 10 g |
Assay Percent Range | 99.8% (metals basis) |
Solubility Information | Soluble in dilute nitric acid. |
Formula Weight | 230.43 (Anhydrous) |
Sensitivity | Hygroscopic |
Appearance | Red-brown cristals |
Chemical Name or Material | Palladium(II) nitrate hydrate |
Safety and Handling
Hazard Category | H272-H290-H302-H314-H335 |
Precautionary Statement | P210-P220-P221-P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P390-P501c |
DOTInformation | DOT Class: 5.1; Packaging Group: II |
EINECSNumber | 233-265-8 |
RTECSNumber | QV0560000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.