Alfa Aesar™ Palladium(II) nitrate, solution, Pd 12-16% w/w (cont. Pd)

Manufacturer: Alfa Aesar™ 41647NH

Quantity 1g

 View more versions of this product

Catalog No. AA41647NH

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10102-05-3
    Formula Weight 230.43
    MDL Number MFCD00011169
    UN Number UN3264
    Chemical Name or Material Palladium(II) nitrate, Solution

    Specifications

    Formula Weight 230.43
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    UN Number UN3264
    RTECS Number QV0560000
    TSCA TSCA
    Color Brown to yellow to orange
    Quantity 1g
    Physical Form Liquid
    MDL Number MFCD00011169
    CAS 10102-05-3
    Health Hazard 1 Danger
    Concentration Pd 12 to 16% w/w (cont. Pd)
    Health Hazard 2 GHS H Statement
    H314-H318-H290-H302-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.
    Chemical Name or Material Palladium(II) nitrate, Solution

    GHS H Statement
    H314-H318-H290-H302-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit