Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Palladium(II) oxide, 86.8-87.1% Pd, Thermo Scientific™
Click to view available options
1g; Glass bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
CAS | 1314-08-5 |
Quantity | 1g |
Molecular Formula | OPd |
Linear Formula | PdO |
MDL Number | MFCD00011172 |
Merck Index | 15, 7094 |
Synonym | palladium ii oxide, palladium oxide pdo, acmc-1bqvh, palladium oxide, pdo, palladium ii oxide, anhydrous, palladium ii oxide-pd, 11113-77-2 cpd with unspecified mf, palladium oxide ? inverted exclamation mark-?, palladium ii oxide trace metals basis |
Solubility Information | Solubility in water: practically insoluble. Other solubilities: practically insoluble in acids, slightly soluble in aqua regia, soluble in 48% hbr |
InChI Key | JQPTYAILLJKUCY-UHFFFAOYSA-N |
SMILES | [O--].[Pd++] |
Show More |
Safety and Handling
EINECSNumber | 215-218-3 |
TSCA | TSCA |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction