Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) oxide, 86.8-87.1% Pd, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1314-08-5
OPd
122.419
MFCD00011172
HBEQXAKJSGXAIQ-UHFFFAOYSA-N
palladium ii oxide,palladium oxide pdo,acmc-1bqvh,palladium oxide, pdo,palladium ii oxide, anhydrous,palladium ii oxide-pd,11113-77-2 cpd with unspecified mf,palladium oxide ? inverted exclamation mark-?,palladium ii oxide trace metals basis
5083724
oxopalladium
O=[Pd]
1314-08-5
OPd
122.419
MFCD00011172
HBEQXAKJSGXAIQ-UHFFFAOYSA-N
palladium ii oxide,palladium oxide pdo,acmc-1bqvh,palladium oxide, pdo,palladium ii oxide, anhydrous,palladium ii oxide-pd,11113-77-2 cpd with unspecified mf,palladium oxide ? inverted exclamation mark-?,palladium ii oxide trace metals basis
5083724
oxopalladium
O=[Pd]
Linear Formula PdO
Merck Index 15, 7094
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in acids, slightly soluble in aqua regia, soluble in 48% hbr
Formula Weight 122.4
Physical Form Powder
Percent Purity 86.8 to 87.1% (Pd)
Density 8.7
Packaging Glass bottle
Solubility Solubility in water: practically insoluble. Other solubilities: practically insoluble in acids, slightly soluble in aqua regia, soluble in 48% hbr
Color Black to Gray
Melting Point 870.0°C
Quantity 5g
Chemical Name or Material Palladium(II) oxide, 86.8-87.1% Pd
Show More Show Less

EINECSNumber : 215-218-3

TSCA : TSCA