Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) oxide, 99.995%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
1314-08-5
OPd
122.4
HBEQXAKJSGXAIQ-UHFFFAOYSA-N
palladium ii oxide, palladium oxide pdo, acmc-1bqvh, palladium oxide, pdo, palladium ii oxide, anhydrous, palladium ii oxide-pd, 11113-77-2 cpd with unspecified mf, palladium oxide ? inverted exclamation mark-?, palladium ii oxide trace metals basis
5083724
oxopalladium
O=[Pd]
1314-08-5
OPd
122.4
HBEQXAKJSGXAIQ-UHFFFAOYSA-N
palladium ii oxide, palladium oxide pdo, acmc-1bqvh, palladium oxide, pdo, palladium ii oxide, anhydrous, palladium ii oxide-pd, 11113-77-2 cpd with unspecified mf, palladium oxide ? inverted exclamation mark-?, palladium ii oxide trace metals basis
5083724
oxopalladium
O=[Pd]
For ultra trace analysis
Assay Percent Range 85.2 to 88.7% (Pd)
Linear Formula PdO
Merck Index 15,7094
Solubility Information Solubility in water: practically insoluble. Other solubilities: practically insoluble in acids,slightly soluble in aqua regia,soluble in 48% hbr
Formula Weight 122.4
Physical Form Viscous Liquid
Percent Purity 99.995%
Grade Trace Metal Basis
Packaging Glass bottle
Solubility Solubility in water: practically insoluble. Other solubilities: practically insoluble in acids, slightly soluble in aqua regia, soluble in 48% hbr
Total Trace Metal Impurities 50ppm max.
Color Undesignated
Melting Point 870.0°C
Quantity 1g
Chemical Name or Material Palladium(II) oxide
Show More Show Less

EINECSNumber : 215-218-3