Alfa Aesar™ Palladium(II) oxide, anhydrous, 99.9% (metals basis), Pd 85% min

Catalytic reduction, in organic synthesis

Manufacturer: Alfa Aesar™ 1104006

Quantity 5g

 View more versions of this product

Catalog No. AA1104006

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1314-08-5
    Formula Weight 122.4
    MDL Number MFCD00011172
    Merck Index 14,6993
    Chemical Name or Material Palladium(II) oxide, Anhydrous

    Specifications

    Formula Weight 122.4
    Odor Odorless
    Melting Point 870°C
    Mesh Size −22 Mesh
    Physical Form Powder
    MDL Number MFCD00011172
    CAS 1314-08-5
    TSCA TSCA
    Merck Index 14,6993
    Color Brown
    Quantity 5g
    Solubility Information Insoluble in water, acids; Slightly soluble in aqua regia
    Density 8.70g/mL
    RTECS Number RT3592900
    Concentration Pd 85% min.
    Chemical Name or Material Palladium(II) oxide, Anhydrous
    Assay 99.9%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit