Alfa Aesar™ Palladium(II) oxide monohydrate, 99.9% (metals basis), Pd 73% min

Manufacturer: Alfa Aesar™ 1104109

Quantity 10g

 View more versions of this product

Catalog No. AA1104109

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 64109-12-2
    Formula Weight 140.41 (122.40 Anhydrous)
    MDL Number MFCD00011172
    Merck Index 14,6993
    Chemical Name or Material Palladium(II) oxide monohydrate

    Specifications

    Formula Weight 140.41 (122.40 Anhydrous)
    Physical Form Powder
    MDL Number MFCD00011172
    Merck Index 14,6993
    TSCA TSCA
    CAS 64109-12-2
    Quantity 10g
    Concentration Pd 73% min.
    Chemical Name or Material Palladium(II) oxide monohydrate
    Assay 99.9%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit