Alfa Aesar™ Palladium(II) sulfate dihydrate, Premion™, 99.995% (metals basis)

Manufacturer: Alfa Aesar™ 4369803

Quantity 1g

 View more versions of this product

Catalog No. AA4369803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13444-98-9
    Molecular Formula O4PdS·2H2O
    Linear Formula PdSO4·2H2O
    Formula Weight 238.51 (202.48 Anhydrous)
    MDL Number MFCD00011173
    UN Number UN3260
    Chemical Name or Material Palladium (II) sulfate dihydrate
    Grade Premion™

    Specifications

    Formula Weight 238.51 (202.48 Anhydrous)
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 2 GHS H Statement
    H314-H318-H290-H302-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.
    May cause an allergic skin reaction.
    Molecular Formula O4PdS·2H2O
    Health Hazard 1 Danger
    Sensitivity Hygroscopic
    MDL Number MFCD00011173
    CAS 13444-98-9
    Quantity 1g
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
    Physical Form Powder
    Linear Formula PdSO4·2H2O
    UN Number UN3260
    Color Red to brown
    Chemical Name or Material Palladium (II) sulfate dihydrate
    Assay 99.995%, (metals basis)
    Grade Premion™

    GHS H Statement
    H314-H318-H290-H302-H317
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    May be corrosive to metals.
    Harmful if swallowed.
    May cause an allergic skin reaction.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit