Palladium Plasma Std. Solution, Alfa Aesar™, Specpure™

Manufacturer: Alfa Aesar 14432AD

Quantity 50mL

 View more versions of this product

Catalog No. AA14432AD

  Check Availability
Add to Cart

    Chemical Identifiers

    Physical Form Liquid
    Molecular Formula Pd in 20% HCl
    MDL Number MFCD03613603
    Chemical Name or Material Palladium

    Specifications

    Quantity 50mL
    DOT Information Hazard Class: 8, Packing Group: II
    EINECS Number 231-595-7
    Physical Form Liquid
    Appearance Liquid
    Molecular Formula Pd in 20% HCl
    MDL Number MFCD03613603
    Health Hazard 1 H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.
    Odor Characteristic
    Chemical Name or Material Palladium

    H314-H335: Causes severe skin burns and eye damage. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit