Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium Powder, Alfa Aesar™

Manufacturer:  Alfa Aesar 1172106

Quantity 5g

 View more versions of this product

Catalog No. AA1172106


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

Powder
MFCD03427451
Palladium, 5% on barium carbonate powder

Specifications

5g
Powder
Odorless
Palladium, 5% on barium carbonate powder
gray powder
MFCD03427451
Gray

Safety and Handling

SDS & Safety Documents

Avoid contact with skin and eyes.

H301 : Toxic if swallowed

DOTInformation : DOT Class: 6.1, Packaging Group: III

Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit