Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium Powder, APS 0.25 - 0.55 micron, 99.95% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 0077677

Quantity 0.1g

 View more versions of this product

Catalog No. AA0077677

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-05-3
99.95% (Metals basis)
Palladium
Powder
MFCD00011167

Specifications

0.1g
Powder
99.95% (Metals basis)
Odorless
7440-05-3
Powder
MFCD00011167
Palladium

Safety and Handling

H228 : Flammable solid

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-115-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit