Alfa Aesar™ Palladium Silver foil, 0.05mm (0.002 in.) thick, 99.9% (metals basis excluding Pt)

Manufacturer: Alfa Aesar™ 42682GH

Quantity 100x100mm

 View more versions of this product

Catalog No. AA42682GH

  Check Availability
Add to Cart

    Chemical Identifiers

    MDL Number MFCD02091738
    Chemical Name or Material Palladium Silver foil, 0.05mm (0.002 in.) thick

    Specifications

    MDL Number MFCD02091738
    TSCA TSCA
    Physical Form Foil
    Odor Odorless
    Avg. Mol. Wt. or Mol. Wt. Range Pd:Ag; 75:25 wt%
    Quantity 100x100mm
    Weight ≈0.36g/25x25mm
    Chemical Name or Material Palladium Silver foil, 0.05mm (0.002 in.) thick
    Assay 99.9%, (metals basis excluding Pt)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit