Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Palladium Silver foil, 0.10mm (0.004 in.) thick, 99.9% (metals basis excluding Pt)

Manufacturer:  Alfa Aesar™ 44731FI

Quantity 50x50mm

 View more versions of this product

Catalog No. AA44731FI

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

MFCD02091738
Palladium Silver foil, 0.10mm (0.004 in.) thick

Specifications

Pd:Ag; 75:25 wt%
50x50mm
MFCD02091738
99.9%, (metals basis excluding Pt)
≈0.72g/25x25mm
Foil
Palladium Silver foil, 0.10mm (0.004 in.) thick

Safety and Handling

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit