Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Palladium Silver foil, 0.5mm (0.02 in.) thick, 99.9% (metals basis excluding Pt)

Manufacturer:  Alfa Aesar™ 42684FF

Quantity 25x25mm

 View more versions of this product

Catalog No. AA42684FF


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

MFCD02091738
Palladium Silver foil, 0.5mm (0.02 in.) thick

Specifications

Pd:Ag; 75:25 wt%
25x25mm
MFCD02091738
Palladium Silver foil, 0.5mm (0.02 in.) thick
≈3.64g/25x25mm
Foil
Odorless
99.9%, (metals basis excluding Pt)

Safety and Handling

SDS & Safety Documents

TSCA : TSCA

missing translation for 'provideContentCorrection'

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

missing translation for 'productTitle'