Palladium Wire, 1mm (0.04 in.) dia., 99.9% (Metals basis), Alfa Aesar™

Form: Approx.9.44g/m

Manufacturer: Alfa Aesar 10280CB

Quantity 250cm

 View more versions of this product

Catalog No. AA10280CB

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-05-3
    Physical Form Wire, 1.0mm (0.04 in.) dia.
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00011167
    Chemical Name or Material Palladium

    Specifications

    Quantity 250cm
    Color Silver-gray
    Physical Form Wire, 1.0mm (0.04 in.) dia.
    Assay Percent Range 99.9% (Metals basis)
    MDL Number MFCD00011167
    CAS 7440-05-3
    Odor Odorless
    Appearance Silver gray wire
    Chemical Name or Material Palladium

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit