Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Palmitoylcarnitine chloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
6865-14-1
C23H46ClNO4
436.074
GAMKNLFIHBMGQT-UHFFFAOYSA-N
palmitoyl-dl-carnitine chloride,+/--palmitoylcarnitine chloride,3-carboxy-n,n,n-trimethyl-2-palmitoyloxy propan-1-aminium chloride,c23h46no4.cl,palmitoylcarnitine chlorid,dl-palmitoyl carnitine hcl,palmityl dl-carnitine chloride,palmitoyl carnitine hydrochloride,o-palmitoyl-dl-carnitine chloride
363417
(3-carboxy-2-hexadecanoyloxypropyl)-trimethylazanium;chloride
CCCCCCCCCCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]
This item is not returnable. View return policy
6865-14-1
C23H46ClNO4
436.074
GAMKNLFIHBMGQT-UHFFFAOYSA-N
palmitoyl-dl-carnitine chloride,+/--palmitoylcarnitine chloride,3-carboxy-n,n,n-trimethyl-2-palmitoyloxy propan-1-aminium chloride,c23h46no4.cl,palmitoylcarnitine chlorid,dl-palmitoyl carnitine hcl,palmityl dl-carnitine chloride,palmitoyl carnitine hydrochloride,o-palmitoyl-dl-carnitine chloride
363417
(3-carboxy-2-hexadecanoyloxypropyl)-trimethylazanium;chloride
CCCCCCCCCCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]

Intermediate in fatty acid oxidation

Intermediate in mitochondrial fatty acid oxidation. Has a wide range of biological actions including the inhibition of protein kinase C and cell membrane disruption.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 436.07
Percent Purity >98%
Chemical Name or Material (±)-Palmitoylcarnitine chloride

Recommended Storage : Desiccate at Room Temperature