Learn More
Selleck Chemical LLCSupplier Diversity Partner Palomid 529 (P529) 50mg 914913-88-5 SG 00529  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S223850MG
Palomid 529 (P529, SG 00529) inhibits both the mTORC1 and mTORC2 complexes, reduces phosphorylation of pAktS473, pGSK3βS9, and pS6. Phase 1.
Catalog No. 50-645-2