Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGAComfort PAM™ 100% Dual Layer Square Memory Foam Insole

Ergonomically designed by podiatrists, Dual layer 100% memory foam, Clinically proven field tested to reduce pain and fatigue, Max shock absorption, Comfort square toe, Western boot friendly

$26.95

Specifications

Color Black, Blue, Yellow
Material Memory Foam
Product Type Specs Square toe insole
Waterproof No
For Use With (Application) Stationary, mobile and working outdoors
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-813-529
View Documents
MEGAComfort
PAMSQW567
Women's: 5-7
Pair for $26.95
Add to cart
 
18-813-530
View Documents
MEGAComfort
PAMSQM67/W89
Men's: 6-7, Women's: 8-9
Pair for $26.95
Add to cart
 
18-813-531
View Documents
MEGAComfort
PAMSQM89/W1011
Men's: 8-9, Women's: 10-11
Pair for $26.95
Add to cart
 
18-813-532
View Documents
MEGAComfort
PAMSQM1011/W1213
Men's: 10-11, Women's: 12-13
Pair for $26.95
Add to cart
 
18-813-534
View Documents
MEGAComfort
PAMSQM1213
Men's: 12-13
Pair for $26.95
Add to cart
 
18-813-535
View Documents
MEGAComfort
PAMSQM1415
Men's: 14-15
Pair for $26.95
Add to cart
 
Description

Description

  • Personal Anti-Fatigue Mat (PAM™) square toe insole
  • Uses patented combination of high-density dual layer 100% memory foam
  • Reduces muscle strain and improves balance reactions
  • Perfect for steel toe footwear as memory foam compresses for safe clearance
  • Ideal for stationary, mobile and working outdoors
  • For use with industrial (safety and steel-toed) and western footwear
  • Contoured arch support for balance, comfort and cushioning
  • Silver ion anti-microbial technology in top cloth offering anti-bacterial, anti-perspiration and anti-odor
Specifications

Specifications

Black, Blue, Yellow
Square toe insole
Stationary, mobile and working outdoors
Memory Foam
No
Industrial (safety and steel-toe) and western footwear
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit