Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGAComfort PAM™ Diabetic Triple Layer Anti-Fatigue Insole

Personal anti-fatigue mat insole ergonomically designed by podiatrists, Dual layer 100% memory foam top layer EVA helps with diabetes, Clinically proven field tested to reduce pain and fatigue, Max shock absorption and comfort

$31.95

Specifications

Color Beige, Blue, Yellow
Material Memory Foam, EVA
Product Type Specs Diabetic insole
Waterproof No
For Use With (Application) Individuals with diabetes mellitus
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
18-813-523
View Documents
MEGAComfort
DIABW567
Women's: 5-7
Pair for $31.95
Add to cart
 
18-813-524
View Documents
MEGAComfort
DIABM67/W89
Men's: 6-7, Women's: 8-9
Pair for $31.95
Add to cart
 
18-813-525
View Documents
MEGAComfort
DIABM89/W1011
Men's: 8-9, Women's: 10-11
Pair for $31.95
Add to cart
 
18-813-526
View Documents
MEGAComfort
DIABM1011/W1213
Men's: 10-11, Women's: 12-13
Pair for $31.95
Add to cart
 
18-813-527
View Documents
MEGAComfort
DIABM1213
Men's: 12-13
Pair for $31.95
Add to cart
 
18-813-528
View Documents
MEGAComfort
DIABM1415
Men's: 14-15
Pair for $31.95
Add to cart
 
Description

Description

  • Personal Anti-Fatigue Mat (PAM™) for diabetics
  • Uses patented combination of high-density dual layer 100% memory foam and includes layer of EVA foam
  • Reduces pain and fatigue while increasing balance and comfort
  • Insole provides max shock absorption to help prevent sheering and comfort in casual footwear
  • Designed for casual footwear due to thickness of insole
  • Ideal for those with diabetes mellitus regardless of any known foot issues
  • Recommended for casual footwear, but can be used with safety footwear

Note:

  • Physician consultation is advised
Specifications

Specifications

Beige, Blue, Yellow
Diabetic insole
Individuals with diabetes mellitus
Memory Foam, EVA
No
Casual and safety footwear
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit